Assay Details
| Gene Symbol: NAA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266880.2 | NP_001253809.1 | 995 | 85 | ||
| GenBank mRNA | JU326896.1 | - | 704 | 85 | ||
| JV046166.1 | - | 995 | 85 | |||
| JV641940.1 | - | 1030 | 85 | |||
More Information
Set Membership: |
|

NAA30
Genomic Map