Assay Details
| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114355.1 | NP_001107827.1 | 1268 | 128 | ||
| GenBank mRNA | AY681964.1 | - | 1268 | 128 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DISC1| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114355.1 | NP_001107827.1 | 1268 | 128 | ||
| GenBank mRNA | AY681964.1 | - | 1268 | 128 | ||
Set Membership: |
|