Assay Details
| Gene Symbol: DISC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114355.1 | NP_001107827.1 | 1792 | 105 | ||
| GenBank mRNA | AY681964.1 | - | 1792 | 105 | ||
| JU330118.1 | - | 68 | 105 | |||
More Information
Set Membership: |
|

DISC1
Genomic Map