Assay Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084767.3 | XP_001084767.3 | 4020 | 136 | ||
| XM_015131318.2 | XP_014986804.2 | 3944 | 136 | |||
More Information
Set Membership: |
|

XIRP1
Genomic Map