Assay Details
Gene Symbol: C2H3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194440.1 | NP_001181369.1 | 645 | 118 | ||
GenBank mRNA | JU323330.1 | - | 611 | 118 | ||
JU323331.1 | - | 611 | 118 | |||
JU473783.1 | - | 636 | 118 | |||
JV045686.1 | - | 652 | 118 | |||
JV567272.1 | - | 643 | 118 | |||
JV639182.1 | - | 654 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
