Assay Details
| Gene Symbol: MRGPRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101665.3 | XP_001101665.2 | 257 | 167 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRGPRD| Gene Symbol: MRGPRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101665.3 | XP_001101665.2 | 257 | 167 | ||
Set Membership: |
|