Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266072.2 | NP_001253001.1 | 769 | 119 | ||
| XM_015134929.2 | XP_014990415.2 | 907 | 119 | |||
| GenBank mRNA | JU321092.1 | - | 699 | 119 | ||
| JU476924.1 | - | 637 | 119 | |||
| JV048024.1 | - | 770 | 119 | |||
More Information
Set Membership: |
|

BPGM
Genomic Map