Assay Details
| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106055.3 | XP_001106055.1 | 954 | 117 | ||
| GenBank mRNA | JU343339.1 | - | 194 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INKA1| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106055.3 | XP_001106055.1 | 954 | 117 | ||
| GenBank mRNA | JU343339.1 | - | 194 | 117 | ||
Set Membership: |
|