Assay Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194690.4 | NP_001181619.1 | 641 | 130 | ||
| GenBank mRNA | JU334450.1 | - | 511 | 130 | ||
| JV667258.1 | - | 645 | 130 | |||
More Information
Set Membership: |
|

RPRML
Genomic Map