Assay Details
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122858.2 | XP_014978344.1 | 1125 | 122 | ||
XM_015122860.2 | XP_014978346.1 | 1159 | 122 | |||
XM_015122861.2 | XP_014978347.1 | 803 | 122 | |||
XM_028839947.1 | XP_028695780.1 | 958 | 122 | |||
GenBank mRNA | JU337339.1 | - | 693 | 122 | ||
JU478041.1 | - | 811 | 122 | |||
JV636762.1 | - | 734 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
