Assay Details
Gene Symbol: C1H1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090172.4 | XP_001090172.1 | 729 | 170 | ||
XM_028849240.1 | XP_028705073.1 | 602 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() |
