Assay Details
| Gene Symbol: SRRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149968.2 | XP_015005454.2 | 193 | 65 | ||
| GenBank mRNA | JU486595.1 | - | 88 | 65 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRRD
Genomic Map