Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194710.1 | NP_001181639.1 | 551 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
