Assay Details
Gene Symbol: LOC709576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107893.4 | XP_001107893.1 | 308 | 126 | ||
GenBank mRNA | JU335309.1 | - | 176 | 126 | ||
JU476268.1 | - | 169 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001107893.4 transcripts.
Set Membership: |
![]() ![]() |
