Assay Details
| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109143.4 | XP_001109143.3 | 711 | 119 | ||
| XM_015150139.2 | XP_015005625.2 | 711 | 119 | |||
| XM_015150140.2 | XP_015005626.2 | 711 | 119 | |||
| XM_028827848.1 | XP_028683681.1 | 620 | 119 | |||
More Information
Set Membership: |
|

CCDC157
Genomic Map