Assay Details
| Gene Symbol: EPM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087462.4 | XP_001087462.1 | 823 | 178 | ||
| XM_028847188.1 | XP_028703021.1 | 267 | 178 | |||
| XM_028847189.1 | XP_028703022.1 | 198 | 178 | |||
| GenBank mRNA | JU326005.1 | - | 399 | 178 | ||
| JU326006.1 | - | 399 | 178 | |||
More Information
Set Membership: |
|

EPM2A
Genomic Map