Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032927.1 | NP_001028099.1 | 280 | 77 | ||
| XM_028834204.1 | XP_028690037.1 | 1219 | 77 | |||
| GenBank mRNA | AY937248.1 | - | 280 | 77 | ||
| JU487500.1 | - | 129 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAEP
Genomic Map