Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134926.2 | XP_014990412.1 | 1410 | 136 | ||
| XM_015134927.2 | XP_014990413.1 | 1438 | 136 | |||
| XM_028846299.1 | XP_028702132.1 | 1200 | 136 | |||
| XM_028846300.1 | XP_028702133.1 | 1333 | 136 | |||
| GenBank mRNA | JV717159.1 | - | 1008 | 136 | ||
More Information
Set Membership: |
|

PODXL
Genomic Map