Assay Details
| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112556.4 | XP_001112556.2 | 341 | 122 | ||
| XM_028828762.1 | XP_028684595.1 | 282 | 122 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPO4| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112556.4 | XP_001112556.2 | 341 | 122 | ||
| XM_028828762.1 | XP_028684595.1 | 282 | 122 | |||
Set Membership: |
|