Assay Details
| Gene Symbol: CTNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089495.4 | XP_001089495.1 | 1278 | 120 | ||
| XM_015118248.2 | XP_014973734.1 | 1294 | 120 | |||
| XM_028835889.1 | XP_028691722.1 | 1341 | 120 | |||
| XM_028835890.1 | XP_028691723.1 | 1153 | 120 | |||
| XM_028835891.1 | XP_028691724.1 | 704 | 120 | |||
| XM_028835892.1 | XP_028691725.1 | 1340 | 120 | |||
| GenBank mRNA | JU323110.1 | - | 120 | |||
| JU472126.1 | - | 120 | ||||
| JV635827.1 | - | 120 | ||||
| JV704164.1 | - | 1004 | 120 | |||
More Information
Set Membership: |
|

CTNS
Genomic Map