Assay Details
| Gene Symbol: RCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257486.1 | NP_001244415.1 | 649 | 64 | ||
| XM_015113535.2 | XP_014969021.1 | 646 | 64 | |||
| XM_015113536.2 | XP_014969022.1 | 457 | 64 | |||
| GenBank mRNA | JU337402.1 | - | 649 | 64 | ||
| JV048868.1 | - | 838 | 64 | |||
More Information
Set Membership: |
|

RCE1
Genomic Map