Assay Details
| Gene Symbol: SLC35A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265836.1 | NP_001252765.1 | 596 | 79 | ||
| XM_015127325.2 | XP_014982811.1 | 697 | 79 | |||
| XM_015127328.2 | XP_014982814.1 | 690 | 79 | |||
| XM_015127329.2 | XP_014982815.1 | 810 | 79 | |||
| GenBank mRNA | JU321333.1 | - | 285 | 79 | ||
| JU321338.1 | - | 285 | 79 | |||
| JU472712.1 | - | 596 | 79 | |||
More Information
Set Membership: |
|

SLC35A2
Genomic Map