Assay Details
| Gene Symbol: CBFA2T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828320.1 | XP_028684153.1 | 1223 | 75 | ||
| XM_028828321.1 | XP_028684154.1 | 1223 | 75 | |||
| XM_028828322.1 | XP_028684155.1 | 1930 | 75 | |||
| GenBank mRNA | JU337054.1 | - | 1063 | 75 | ||
| JU477782.1 | - | 1165 | 75 | |||
| JV665276.1 | - | 1075 | 75 | |||
More Information
Set Membership: |
|

CBFA2T2
Genomic Map