Assay Details
| Gene Symbol: EXTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267005.1 | NP_001253934.1 | 694 | 138 | ||
| GenBank mRNA | JU322414.1 | - | 802 | 138 | ||
| JU473155.1 | - | 738 | 138 | |||
| JV043755.1 | - | 694 | 138 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EXTL2
Genomic Map