Assay Details
| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125559.2 | XP_014981045.1 | 1183 | 69 | ||
| XM_028849925.1 | XP_028705758.1 | 1378 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMX1A| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125559.2 | XP_014981045.1 | 1183 | 69 | ||
| XM_028849925.1 | XP_028705758.1 | 1378 | 69 | |||
Set Membership: |
|