Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257731.1 | NP_001244660.1 | 536 | 119 | ||
| XM_015149642.2 | XP_015005128.1 | 581 | 119 | |||
| GenBank mRNA | JU319921.1 | - | 536 | 119 | ||
| JV722554.1 | - | 552 | 119 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UFD1
Genomic Map