Assay Details
| Gene Symbol: LOC114669817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842712.1 | XP_028698545.1 | 1117 | 155 | ||
| Gene Symbol: CEP250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149122.2 | XP_015004608.1 | 1202 | 155 | ||
| XM_028828364.1 | XP_028684197.1 | 1203 | 155 | |||
| XM_028828368.1 | XP_028684201.1 | 1076 | 155 | |||
| GenBank mRNA | JU332731.1 | - | 1081 | 155 | ||
| JU474154.1 | - | 1086 | 155 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028828368.1 transcripts.
Set Membership: |
|

LOC114669817
Genomic Map