Assay Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118637.2 | XP_014974123.1 | 1281 | 102 | ||
| XM_028836032.1 | XP_028691865.1 | 900 | 102 | |||
| XM_028836033.1 | XP_028691866.1 | 562 | 102 | |||
| XM_028836034.1 | XP_028691867.1 | 1774 | 102 | |||
| XM_028836035.1 | XP_028691868.1 | 322 | 102 | |||
| GenBank mRNA | JV049913.1 | - | 313 | 102 | ||
| JV640451.1 | - | 953 | 102 | |||
More Information
Set Membership: |
|

TRIM16
Genomic Map