Assay Details
| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098858.4 | XP_001098858.2 | 467 | 139 | ||
| XM_028831152.1 | XP_028686985.1 | 467 | 139 | |||
| GenBank mRNA | JU480354.1 | - | 183 | 139 | ||
More Information
Set Membership: |
|

AP1S3
Genomic Map