Assay Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028833109.1 | XP_028688942.1 | 668 | 126 | ||
| XM_028833110.1 | XP_028688943.1 | 668 | 126 | |||
| XM_028833111.1 | XP_028688944.1 | 668 | 126 | |||
| XM_028833112.1 | XP_028688945.1 | 668 | 126 | |||
| GenBank mRNA | JU496814.1 | - | 602 | 126 | ||
More Information
Set Membership: |
|

TESMIN
Genomic Map