Assay Details
| Gene Symbol: LOC697753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088873.4 | XP_001088873.2 | 363 | 100 | ||
| XM_015146529.2 | XP_015002015.2 | 363 | 100 | |||
| XM_028826008.1 | XP_028681841.1 | 247 | 100 | |||
| GenBank mRNA | JU323595.1 | - | 235 | 100 | ||
| JV718168.1 | - | 241 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001088873.4, XM_015146529.2, XM_028826008.1 transcripts.
Set Membership: |
|

LOC697753
Genomic Map