Assay Details
| Gene Symbol: LOC704512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127518.2 | XP_014983004.1 | 1835 | 125 | ||
| XM_015127519.2 | XP_014983005.1 | 1820 | 125 | |||
| XM_015127520.1 | XP_014983006.1 | 1641 | 125 | |||
| XM_015127521.1 | XP_014983007.1 | 1575 | 125 | |||
| XM_015127522.1 | XP_014983008.1 | 1622 | 125 | |||
| XM_015127523.1 | XP_014983009.1 | 1548 | 125 | |||
| XM_015127524.1 | XP_014983010.1 | 1548 | 125 | |||
| XM_015127525.2 | XP_014983011.2 | 644 | 125 | |||
| XM_015127527.2 | XP_014983013.1 | 629 | 125 | |||
| XM_028842516.1 | XP_028698349.1 | 2050 | 125 | |||
| XM_028842517.1 | XP_028698350.1 | 1653 | 125 | |||
| XM_028842518.1 | XP_028698351.1 | 462 | 125 | |||
| GenBank mRNA | JU477031.1 | - | 1513 | 125 | ||
| JV592381.1 | - | 1594 | 125 | |||
More Information
Set Membership: |
|

LOC704512
Genomic Map