Assay Details
Gene Symbol: ZNF384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002798463.3 | XP_002798509.1 | 1467 | 72 | ||
XM_015150918.2 | XP_015006404.1 | 1938 | 72 | |||
XM_015150919.2 | XP_015006405.1 | 1919 | 72 | |||
XM_015150921.2 | XP_015006407.1 | 1536 | 72 | |||
XM_015150923.2 | XP_015006409.1 | 1546 | 72 | |||
XM_015150924.2 | XP_015006410.1 | 1695 | 72 | |||
XM_015150925.2 | XP_015006411.1 | 1488 | 72 | |||
XM_015150926.2 | XP_015006412.1 | 1443 | 72 | |||
XM_015150927.2 | XP_015006413.1 | 1602 | 72 | |||
XM_028829205.1 | XP_028685038.1 | 1448 | 72 | |||
XM_028829206.1 | XP_028685039.1 | 1701 | 72 | |||
XM_028829208.1 | XP_028685041.1 | 1871 | 72 | |||
XM_028829209.1 | XP_028685042.1 | 1823 | 72 | |||
XM_028829210.1 | XP_028685043.1 | 1775 | 72 | |||
XM_028829211.1 | XP_028685044.1 | 1736 | 72 | |||
XM_028829212.1 | XP_028685045.1 | 1640 | 72 | |||
XM_028829214.1 | XP_028685047.1 | 1592 | 72 | |||
GenBank mRNA | JU474534.1 | - | 1355 | 72 | ||
JU474535.1 | - | 1172 | 72 | |||
JU474536.1 | - | 1124 | 72 | |||
JU474537.1 | - | 1490 | 72 | |||
JV046335.1 | - | 1338 | 72 | |||
JV046336.1 | - | 1677 | 72 | |||
JV046337.1 | - | 1386 | 72 | |||
JV046338.1 | - | 1203 | 72 | |||
JV635992.1 | - | 1348 | 72 | |||
JV635993.1 | - | 1165 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
