Assay Details
| Gene Symbol: LOC694628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146952.2 | XP_015002438.1 | 1226 | 78 | ||
| XM_028826374.1 | XP_028682207.1 | 1130 | 78 | |||
More Information
Set Membership: |
|

LOC694628
Genomic Map