Assay Details
| Gene Symbol: PHF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121078.2 | XP_014976564.1 | 1029 | 85 | ||
| XM_015121079.2 | XP_014976565.1 | 864 | 85 | |||
| XM_015121081.2 | XP_014976567.1 | 4524 | 85 | |||
| XM_015121082.2 | XP_014976568.2 | 1254 | 85 | |||
| XM_028837174.1 | XP_028693007.1 | 460 | 85 | |||
| GenBank mRNA | JU334801.1 | - | 667 | 85 | ||
| JU475829.1 | - | 673 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PHF11
Genomic Map