Assay Details
| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099611.4 | XP_001099611.2 | 552 | 109 | ||
| XM_015111823.2 | XP_014967309.2 | 553 | 109 | |||
| XM_015111824.2 | XP_014967310.2 | 391 | 109 | |||
| XM_028831936.1 | XP_028687769.1 | 553 | 109 | |||
More Information
Set Membership: |
|

FAM228A
Genomic Map