Assay Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260556.1 | NP_001247485.1 | 1298 | 70 | ||
XM_015126325.2 | XP_014981811.1 | 1000 | 70 | |||
XM_015126326.2 | XP_014981812.1 | 478 | 70 | |||
XM_028840393.1 | XP_028696226.1 | 1420 | 70 | |||
GenBank mRNA | JU326747.1 | - | 574 | 70 | ||
JV047980.1 | - | 1298 | 70 | |||
JV047981.1 | - | 1298 | 70 | |||
JV635406.1 | - | 1517 | 70 |

More Information
Set Membership: |
![]() ![]() |
