Assay Details
| Gene Symbol: KIAA0895L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260556.1 | NP_001247485.1 | 1605 | 147 | ||
| XM_015126325.2 | XP_014981811.1 | 1307 | 147 | |||
| XM_015126326.2 | XP_014981812.1 | 785 | 147 | |||
| XM_028840393.1 | XP_028696226.1 | 1727 | 147 | |||
| GenBank mRNA | JU326747.1 | - | 881 | 147 | ||
| JV047980.1 | - | 1605 | 147 | |||
| JV047981.1 | - | 1605 | 147 | |||
| JV635406.1 | - | 1824 | 147 | |||
More Information
Set Membership: |
|

KIAA0895L
Genomic Map