Assay Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093604.4 | XP_001093604.1 | 663 | 87 | ||
| XM_015142681.2 | XP_014998167.1 | 728 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

REC114| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093604.4 | XP_001093604.1 | 663 | 87 | ||
| XM_015142681.2 | XP_014998167.1 | 728 | 87 | |||
Set Membership: |
|