Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121160.2 | XP_014976646.2 | 1827 | 123 | ||
| XM_015121161.2 | XP_014976647.1 | 2100 | 123 | |||
| XM_015121168.2 | XP_014976654.1 | 2064 | 123 | |||
| XM_015121172.2 | XP_014976658.1 | 1617 | 123 | |||
| GenBank mRNA | JU320437.1 | - | 1829 | 123 | ||
| JU476939.1 | - | 1644 | 123 | |||
| JU476940.1 | - | 1822 | 123 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RO60
Genomic Map