Assay Details
| Gene Symbol: VCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107924.4 | XP_001107924.1 | 423 | 129 | ||
| GenBank mRNA | JU477079.1 | - | 163 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VCAM1| Gene Symbol: VCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107924.4 | XP_001107924.1 | 423 | 129 | ||
| GenBank mRNA | JU477079.1 | - | 163 | 129 | ||
Set Membership: |
|