Assay Details
| Gene Symbol: SERPINA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195350.2 | NP_001182279.2 | 1116 | 141 | ||
| XM_015144281.2 | XP_014999767.2 | 1175 | 141 | |||
| XM_015144282.2 | XP_014999768.2 | 1133 | 141 | |||
| GenBank mRNA | JV641848.1 | - | 1114 | 141 | ||
More Information
Set Membership: |
|

SERPINA3
Genomic Map