Assay Details
| Gene Symbol: AANAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047127.1 | NP_001040592.1 | 225 | 144 | ||
| GenBank mRNA | U46661.2 | - | 225 | 144 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AANAT
Genomic Map