Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100020.4 | XP_001100020.1 | 739 | 82 | ||
| XM_028835103.1 | XP_028690936.1 | 910 | 82 | |||
| XM_028835104.1 | XP_028690937.1 | 910 | 82 | |||
| XM_028835105.1 | XP_028690938.1 | 897 | 82 | |||
| XM_028835106.1 | XP_028690939.1 | 790 | 82 | |||
| XM_028835107.1 | XP_028690940.1 | 739 | 82 | |||
| GenBank mRNA | JV650535.1 | - | 146 | 82 | ||
More Information
Set Membership: |
|

AQP7
Genomic Map