Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110720.2 | XP_014966206.2 | 1341 | 96 | ||
| GenBank mRNA | JU494810.1 | - | 206 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASP10| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110720.2 | XP_014966206.2 | 1341 | 96 | ||
| GenBank mRNA | JU494810.1 | - | 206 | 96 | ||
Set Membership: |
|