Assay Details
| Gene Symbol: CNGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101944.4 | XP_001101944.2 | 821 | 87 | ||
| XM_015112928.2 | XP_014968414.2 | 2653 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNGA3| Gene Symbol: CNGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101944.4 | XP_001101944.2 | 821 | 87 | ||
| XM_015112928.2 | XP_014968414.2 | 2653 | 87 | |||
Set Membership: |
|