Assay Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106814.4 | XP_001106814.3 | 504 | 114 | ||
| XM_015118134.2 | XP_014973620.2 | 458 | 114 | |||
| XM_028835374.1 | XP_028691207.1 | 392 | 114 | |||
| XM_028835375.1 | XP_028691208.1 | 398 | 114 | |||
| XM_028835376.1 | XP_028691209.1 | 456 | 114 | |||
| XM_028835377.1 | XP_028691210.1 | 405 | 114 | |||
| GenBank mRNA | JU323374.1 | - | 318 | 114 | ||
| JU481076.1 | - | 255 | 114 | |||
| JV044398.1 | - | 291 | 114 | |||
| JV637693.1 | - | 318 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CTSV
Genomic Map