Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099090.4 | XP_001099090.1 | 1349 | 69 | ||
| XM_028836740.1 | XP_028692573.1 | 1349 | 69 | |||
More Information
Set Membership: |
|

HCRTR1
Genomic Map