Assay Details
| Gene Symbol: CCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144252.1 | XP_014999738.1 | 1092 | 140 | ||
| GenBank mRNA | JV635520.1 | - | 1075 | 140 | ||
| JV713972.1 | - | 742 | 140 | |||
More Information
Set Membership: |
|

CCN1
Genomic Map