Assay Details
| Gene Symbol: BAAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002800027.3 | XP_002800073.2 | 746 | 77 | ||
| GenBank mRNA | CO583221.1 | - | 574 | 77 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BAAT
Genomic Map